EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact dermatitis due to diethyl sebacate.
PMID 9928807
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
246077
Diethyl sebacate, 98%
C14H26O4
W237604
Diethyl sebacate, ≥98%, FCC, FG
C14H26O4