EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A Readily Available and User-Friendly Chiral Catalyst for Efficient Enantioselective Olefin Metathesis This research was supported by the National Institutes of Health (GM-59426) and the National Science Foundation (CHE-9905806 to A. H. H. and CHE-9988766 to R. R. S.).
PMID 11317300
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5644
(±)-Methoxyverapamil hydrochloride, ≥98%
C28H40N2O5 · HCl