EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
The synthesis and biological activity of a series of 2,4-diaminopyrido[2,3-d]pyrimidine based antifolates as antineoplastic and antiarthritic agents.
PMID 9990460
Abstract
A new series of 2,4-diaminopyrido[2,3-d]pyrimidine based antifolates 1-3 were synthesized through an efficient conversion of 2-pivaloyl-4-oxo-6-ethynylpyrido[2,3-d]pyrimidine 5 to the corresponding 4-amino analog 7 via the activated 1,2,4-triazole intermediate 6. Compound 7 was used as the key intermediate for the preparation of the final products. The detailed biological evaluation of these compounds both as antineoplastic and antiarthritic agents will be discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0040
Lometrexol hydrate, ≥95% (HPLC)
C21H25N5O6 · xH2O