EMAIL THIS PAGE TO A FRIEND
115649 Sigma-Aldrich

Piperonylonitrile

97%

Synonym: 3,4-(Methylenedioxy)benzonitrile

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds - Organic Building Blocks,
Quality Level   100
assay   97%
mp   91-93 °C (lit.)
SMILES string   N#Cc1ccc2OCOc2c1
InChI   1S/C8H5NO2/c9-4-6-1-2-7-8(3-6)11-5-10-7/h1-3H,5H2
InChI key   PKRWWZCDLJSJIF-UHFFFAOYSA-N

Description

Application

Piperonylonitrile has been used to alter the growth of Nif + and Nif - strains of K.pneumoniae.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
TO2645000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?