• USA Home
  • 122955 - 4-Aminohippuric acid

EMAIL THIS PAGE TO A FRIEND
122955 Aldrich

4-Aminohippuric acid

97%

Synonym: 4-Aminohippuric acid, N-(4-Aminobenzoyl)glycine, p-Aminohippuric acid

  • CAS Number 61-78-9

  • Linear Formula NH2C6H4C(O)NHCH2COOH

  • Molecular Weight 194.19

  •  Beilstein Registry Number 1213676

  •  EC Number 200-518-9

  •  MDL number MFCD00007890

Purchase

Properties

InChI Key   HSMNQINEKMPTIC-UHFFFAOYSA-N
assay   97%
mp   199-200 °C (dec.) (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A1422 p-Aminohippuric acid, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?