• USA Home
  • 130931 - 2-Chlorobenzylidenemalononitrile

EMAIL THIS PAGE TO A FRIEND
130931 Aldrich

2-Chlorobenzylidenemalononitrile

96%

Purchase

Properties

assay   96%
mp   93-97 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3448 6.1 / PGII
WGK Germany 
3
RTECS 
OO3675000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?