EMAIL THIS PAGE TO A FRIEND
133086 Sigma-Aldrich

2-Methyl-1-propanol

99%

Synonym: Isobutanol, Isobutyl alcohol

  • CAS Number 78-83-1

  • Linear Formula (CH3)2CHCH2OH

  • Molecular Weight 74.12

  •  Beilstein/REAXYS Number 1730878

  •  EC Number 201-148-0

  •  MDL number MFCD00004740

  •  PubChem Substance ID 329750132

Purchase

Properties

vapor density   2.55 (vs air)
vapor pressure   8 mmHg ( 20 °C)
  8.8 mmHg ( 0 °C)
assay   99%
autoignition temp.   801 °F
expl. lim.   10.6 %
refractive index   n20/D 1.396 (lit.)
bp   108 °C (lit.)
  108 °C
mp   −108 °C (lit.)
density   0.803 g/mL at 25 °C (lit.)
SMILES string   CC(C)CO
InChI   1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3
InChI key   ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1212 - class 3 - PG 3 - Isobutanol
WGK Germany 
1
RTECS 
NP9625000
Flash Point(F) 
82.4 °F
Flash Point(C) 
28 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?