EMAIL THIS PAGE TO A FRIEND
134538 Sigma-Aldrich

Boc-Gly-OH

98%

Synonym: N-(tert-Butoxycarbonyl)glycine, Boc-glycine

  • CAS Number 4530-20-5

  • Linear Formula (CH3)3COCONHCH2COOH

  • Molecular Weight 175.18

  •  Beilstein/REAXYS Number 1101514

  •  EC Number 224-864-5

  •  MDL number MFCD00002690

  •  PubChem Substance ID 24848162

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Glycine, Peptide Synthesis and Peptide Chemistry More...
InChI Key   VRPJIFMKZZEXLR-UHFFFAOYSA-N
assay   98%
mp   86-89 °C (lit.)

Description

Other Notes

This product has been replaced by 15420-ALDRICH | Boc-Gly-OH ≥99.0% (T)

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?