EMAIL THIS PAGE TO A FRIEND
143642 Sigma-Aldrich

Homovanillic acid

98%

Synonym: 4-Hydroxy-3-methoxyphenylacetic acid, HVA

  • CAS Number 306-08-1

  • Linear Formula HOC6H3(OCH3)CH2CO2H

  • Molecular Weight 182.17

  •  Beilstein/REAXYS Number 2213447

  •  EC Number 206-176-7

  •  MDL number MFCD00004350

  •  PubChem Substance ID 329750273

Purchase

Properties

assay   98%
mp   142-145 °C (lit.)
SMILES string   COc1cc(CC(O)=O)ccc1O
InChI   1S/C9H10O4/c1-13-8-4-6(5-9(11)12)2-3-7(8)10/h2-4,10H,5H2,1H3,(H,11,12)
InChI key   QRMZSPFSDQBLIX-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis Report of Organic Acids on Discovery® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis Report of Organic Acids on Discovery® C18
Keywords: Chromatography, High performance liquid chromatography

HPLC Analysis of Organic Acids on Discovery® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Organic Acids on Discovery® C18
Keywords: Chromatography, High performance liquid chromatography

Related Products

replaced by

Product #

Description

Add to Cart

H1252 Homovanillic acid, Fluorimetric reagent

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?