EMAIL THIS PAGE TO A FRIEND
151351 Sigma-Aldrich

2,5-Lutidine

95%

Synonym: 2,5-Dimethylpyridine

Purchase

Properties

Related Categories Building Blocks, C7, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
Quality Level   100
vapor pressure   2.4 mmHg ( 20 °C)
assay   95%
refractive index   n20/D 1.499 (lit.)
bp   157 °C (lit.)
mp   −15 °C (lit.)
density   0.926 g/mL at 25 °C (lit.)
SMILES string   Cc1ccc(C)nc1
InChI   1S/C7H9N/c1-6-3-4-7(2)8-5-6/h3-5H,1-2H3
InChI key   XWKFPIODWVPXLX-UHFFFAOYSA-N

Description

General description

The FT-IR and FT-Raman spectra of 2,5-lutidine was studied.

Application

2,5-Lutidine was used to displace CO from the haem iron.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
RTECS 
OK9625000
Flash Point(F) 
118.4 °F
Flash Point(C) 
48 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?