• USA Home
  • 172820 - 1,1,2-Trichlorotrifluoroethane

EMAIL THIS PAGE TO A FRIEND
172820 Sigma-Aldrich

1,1,2-Trichlorotrifluoroethane

ReagentPlus®, 99%

Synonym: 1,1,2-Trichloro-1,2,2-trifluoroethane, 1,1,2-Trichloro-trifluoroethane, Genetron® 113

  • CAS Number 76-13-1

  • Linear Formula ClCF2CCl2F

  • Molecular Weight 187.38

  •  Beilstein/REAXYS Number 1740335

  •  EC Number 200-936-1

  •  MDL number MFCD00000782

  •  PubChem Substance ID 329751299

Purchase

Properties

vapor density   6.5 (vs air)
vapor pressure   5.5 psi ( 20 °C)
product line   ReagentPlus®
assay   99%
autoignition temp.   1256 °F
refractive index   n20/D 1.358 (lit.)
bp   47-48 °C (lit.)
mp   −35 °C (lit.)
density   1.57 g/mL at 25 °C (lit.)
SMILES string   FC(F)(Cl)C(F)(Cl)Cl
InChI   1S/C2Cl3F3/c3-1(4,6)2(5,7)8
InChI key   AJDIZQLSFPQPEY-UHFFFAOYSA-N

Description

Application

Please view www.aldrich.com/epaods regarding the EPA′s request for application information of Ozone Depleting Substances

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Genetron is a registered trademark of Honeywell International Inc.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
WGK 2
RTECS 
KJ4000000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?