EMAIL THIS PAGE TO A FRIEND
178217 Sigma-Aldrich

Ethyl oxanilate

98%

Purchase

Properties

assay   98%
mp   67-69 °C (lit.)
SMILES string   CCOC(=O)C(=O)Nc1ccccc1
InChI   1S/C10H11NO3/c1-2-14-10(13)9(12)11-8-6-4-3-5-7-8/h3-7H,2H2,1H3,(H,11,12)
InChI key   YDGAUBHNAKCSKF-UHFFFAOYSA-N

Description

Application

Ethyl oxanilate derivatives were used as gametocides on the hermaphrodite plants (eg. common wheat).

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?