• USA Home
  • 193011 - 2-Furaldehyde diethyl acetal

EMAIL THIS PAGE TO A FRIEND
193011 Sigma-Aldrich

2-Furaldehyde diethyl acetal

97%

Purchase

Properties

Related Categories Building Blocks, C8 to C11, Chemical Synthesis, Furans, Heterocyclic Building Blocks More...
assay   97%
refractive index   n20/D 1.444 (lit.)
bp   189-191 °C (lit.)
density   1.008 g/mL at 25 °C (lit.)
SMILES string   CCOC(OCC)c1ccco1
InChI   1S/C9H14O3/c1-3-10-9(11-4-2)8-6-5-7-12-8/h5-7,9H,3-4H2,1-2H3
InChI key   SEILDMUKBMYIEZ-UHFFFAOYSA-N

Description

Application

2-Furaldehyde diethyl acetal was used in the synthesis of:
• 5-pyridyl- and 5-aryl-2-furaldehydes via Palladium-mediated cross-coupling reaction
• 11-oxatricyclo [5.3. 1.02, 6] undecane derivatives

Packaging

5, 25 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?