• USA Home
  • 193216 - 2-Chloronicotinamide

EMAIL THIS PAGE TO A FRIEND
193216 Sigma-Aldrich

2-Chloronicotinamide

98%

Purchase

Properties

Related Categories Building Blocks, C6, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
Quality Level   100
assay   98%
mp   164-167 °C (lit.)
SMILES string   NC(=O)c1cccnc1Cl
InChI   1S/C6H5ClN2O/c7-5-4(6(8)10)2-1-3-9-5/h1-3H,(H2,8,10)
InChI key   ZQZAHPFFZWEUCL-UHFFFAOYSA-N

Description

Application

2-Chloronicotinamide was used in the synthesis of pyrrolopyridines and thiol ester derived from 2-mecaptonicotinamide.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?