• USA Home
  • 203831 - Neodymium(III) chloride hexahydrate

EMAIL THIS PAGE TO A FRIEND
203831 Aldrich

Neodymium(III) chloride hexahydrate

99.99%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Materials Science, Metal and Ceramic Science,
InChI Key   PCORICZHGNSNIZ-UHFFFAOYSA-K
assay   99.99%
mp   124 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?