EMAIL THIS PAGE TO A FRIEND
219770 Sigma-Aldrich

DL-Homoserine

99%

Synonym: (±)-2-Amino-4-hydroxybutyric acid

Purchase

Properties

assay   99%
mp   188-189 °C (lit.)
SMILES string   NC(CCO)C(O)=O
InChI   1S/C4H9NO3/c5-3(1-2-6)4(7)8/h3,6H,1-2,5H2,(H,7,8)
InChI key   UKAUYVFTDYCKQA-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?