• USA Home
  • 230979 - 3′,3′′,5′,5′′-Tetraiodophenolsulfonephthalein

EMAIL THIS PAGE TO A FRIEND
230979 Aldrich

3′,3′′,5′,5′′-Tetraiodophenolsulfonephthalein

Dye content 90 %

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes, T-U-V More...
InChI Key   RXNYILISXGUYFG-UHFFFAOYSA-N
composition   Dye content, 90%
mp   229 °C (dec.)(lit.)
absorption   λmax 433 nm

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Milli-Q® Water Purification Solutions

Biomedical Applications
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?