EMAIL THIS PAGE TO A FRIEND
239976 Sigma-Aldrich

Cyanoacetic acid

≥99%

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
vapor pressure   0.1 mmHg ( 100 °C)
InChI Key   MLIREBYILWEBDM-UHFFFAOYSA-N
assay   ≥99%
bp   108 °C/0.15 mmHg (lit.)
mp   64-70 °C (lit.)
solubility   alcohol: soluble(lit.)
  benzene: slightly soluble(lit.)
  chloroform: slightly soluble(lit.)
  diethyl ether: soluble(lit.)
  water: soluble(lit.)

Description

Application

Cyanoacetic acid was used in the synthesis of N-piperidine-cyanacetamide and N-morpholyl-cyanacetamide. It was also used in the preparation of a panchromatic dye for dye-sensitized solar cells.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates toxic gas.
RIDADR 
UN 3261 8 / PGII
WGK Germany 
1
RTECS 
AG3675000
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?