• USA Home
  • 255769 - Thallium(I) carbonate

EMAIL THIS PAGE TO A FRIEND
255769 Aldrich

Thallium(I) carbonate

≥99.99% trace metals basis

Synonym: Thallous carbonate

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Materials Science, Metal and Ceramic Science,
InChI Key   DASUJKKKKGHFBF-UHFFFAOYSA-L
assay   ≥99.99% trace metals basis
form   powder
mp   260-270 °C (dec.) (lit.)
solubility   ethanol: insoluble(lit.)
density   7.11 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1707 6.1 / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?