EMAIL THIS PAGE TO A FRIEND
266752 Sigma-Aldrich

Hafnium

powder, −325 mesh, 99.5% trace metals basis (purity excludes ~2% zirconium), contains 1:10 pentanol to water solution as stabilizer

Purchase

Properties

Related Categories 3D Printing Materials for Research and Development, Hafnium, Materials Science, Metal and Ceramic Science, Metals,
InChI Key   VBJZVLUMGGDVMO-UHFFFAOYSA-N
assay   99.5% trace metals basis (purity excludes ~2% zirconium)
form   powder
contains   1:10 pentanol to water solution as stabilizer
resistivity   29.6 μΩ-cm, 0°C
particle size   −325 mesh
bp   4602 °C(lit.)
mp   2227 °C (lit.)
density   13.3 g/cm3(lit.)

Description

Packaging

25 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1326 - class 4.1 - PG 2 - Hafnium powder, wetted
WGK Germany 
nwg
Protocols & Articles

Articles

3D Printing of Carbon Fiber-Reinforced Composites

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries, most typically using a layer-by-layer fabrication process. 3D pri...
Zhenyu Bo* (Ph.D Candidate at Northwestern University) and Jia Choi*, PhD, Product Manager


*Materials Science Product Management Team, MilliporeSigma, Milwaukee, WI.
Keywords: Deposition, Nanomaterials, Nanotubes

Additive Manufacturing of Permanent Magnets

M. P. Paranthaman,1 I. C. Nlebedim,2 F. Johnson,3 S. K. McCall4 1Oak Ridge National Laboratory, Oak Ridge, TN 37831, USA 2Ames Laboratory, Ames, IA 50011, USA 3GE Global Research, Niskayuna, NY 12309...
M. P. Paranthaman, I. C. Nlebedim, F. Johnson, S. K. McCall
Material Matters, 2016, 11.4
Keywords: Degradations, Deposition, Melting, Nucleic acid annealing, Renewable energy

Self-Propagating Reactions Induced by Mechanical Alloying

Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill,...
Prof. Laszlo Takacs
Material Matters 2007, 2.4, 21.
Keywords: Ceramics, Chemical reactions, Combustion, Diffraction, Exothermic, Mass spectrometry, Metal Science, Oxidations, Reductions, Size-exclusion chromatography, Thermite, X-Ray diffraction, transformation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?