EMAIL THIS PAGE TO A FRIEND
268186 Aldrich

cis-1,3-Pentadiene

98%

Synonym: cis-1-Methyl-1,3-butadiene, cis-Piperylene

Purchase

Properties

vapor pressure   6.58 psi ( 20 °C)
InChI Key   PMJHHCWVYXUKFD-PLNGDYQASA-N
assay   98%
refractive index   n20/D 1.436(lit.)
  n20/D 1.437(lit.)
bp   44 °C(lit.)
mp   −141 °C (lit.)
density   0.694 g/mL at 20 °C
  0.691 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3295BE 3 / PGII
WGK Germany 
3
Flash Point(F) 
5 °F
Flash Point(C) 
-15 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?