• USA Home
  • 270911 - Sodium cholate hydrate

EMAIL THIS PAGE TO A FRIEND
270911 Sigma-Aldrich

Sodium cholate hydrate

98%

Synonym: 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt, Cholalic acid sodium salt

  • CAS Number 206986-87-0

  • Empirical Formula (Hill Notation) C24H39NaO5 · xH2O

  • Molecular Weight 430.55 (anhydrous basis)

  •  Beilstein Registry Number 3582354

  •  EC Number 206-643-5

  •  MDL number MFCD00150749

Purchase

Properties

InChI Key   MUVVIYFKOVLQHL-RCVKHMDESA-M
assay   98%
optical activity   [α]20/D +31.3°, c = 0.5 in H2O
mol wt   micellar avg mol wt 900-1300
aggregation number   2-3
CMC   9-15 mM(20-25°C)
HLB   18

Description

Application

Non-denaturing detergent used for extraction of membrane proteins.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Manufacturing, Characterization and Use of Single Walled Carbon Nanotubes

Carbon nanotubes are materials that possess remarkable properties and offer extraordinary possibilities. This article gives a brief overview of the physico-chemical nature and characterization of sin...
Richard Jansen, Philip Wallis
Material Matters 2009, 4.1, 23.
Keywords: Absorption, Catalysis, Centrifugation, Chemical vapor deposition, Diffraction, Indicators, Infrared spectroscopy, Nanotechnology, Nanotubes, Oxidations, Purification, Raman spectroscopy, Semiconductor, Solar cells, Sonication, Spectroscopy, Ultraviolet-Visible spectroscopy, X-Ray diffraction

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?