• USA Home
  • 275646 - γ-Oxo-1-pyrenebutyric acid

EMAIL THIS PAGE TO A FRIEND
275646 Sigma-Aldrich

γ-Oxo-1-pyrenebutyric acid

technical grade

  • CAS Number 7499-60-7

  • Empirical Formula (Hill Notation) C20H14O3

  • Molecular Weight 302.32

  •  MDL number MFCD00010369

  •  PubChem Substance ID 24856661

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C13 to C42+, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
Quality Level   100
grade   technical grade
mp   184 °C (dec.) (lit.)
SMILES string   OC(=O)CCC(=O)c1ccc2ccc3cccc4ccc1c2c34
InChI   1S/C20H14O3/c21-17(10-11-18(22)23)15-8-6-14-5-4-12-2-1-3-13-7-9-16(15)20(14)19(12)13/h1-9H,10-11H2,(H,22,23)
InChI key   AAJQTWWVKREOQP-UHFFFAOYSA-N

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?