• USA Home
  • 282715 - 3,4-Pyridinedicarboxylic anhydride

EMAIL THIS PAGE TO A FRIEND
282715 Sigma-Aldrich

3,4-Pyridinedicarboxylic anhydride

97%

Synonym: Cinchomeronic anhydride

  • CAS Number 4664-08-8

  • Empirical Formula (Hill Notation) C7H3NO3

  • Molecular Weight 149.10

  •  MDL number MFCD00010680

  •  PubChem Substance ID 24857065

  •  NACRES NA.23

Purchase

Properties

Related Categories Anhydride Monomers, Materials Science, Monomers, Polymer Science
Quality Level   100
assay   97%
mp   75-77 °C (lit.)
SMILES string   O=C1OC(=O)c2cnccc12
InChI   1S/C7H3NO3/c9-6-4-1-2-8-3-5(4)7(10)11-6/h1-3H
InChI key   KFKMGUPDWTWQFM-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?