• USA Home
  • 285013 - (S)-(−)-2-Amino-4-pentenoic acid

EMAIL THIS PAGE TO A FRIEND
285013 Sigma-Aldrich

(S)-(−)-2-Amino-4-pentenoic acid

98%

Synonym: L-2-Amino-4-pentenoic acid, L-Allylglycine

  • CAS Number 16338-48-0

  • Linear Formula H2C=CHCH2CH(NH2)CO2H

  • Molecular Weight 115.13

  •  Beilstein/REAXYS Number 1721512

  •  MDL number MFCD00002627

  •  PubChem Substance ID 24856862

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Linear Core Amino Acids, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
assay   98%
optical activity   [α]20/D −36°, c = 4 in H2O
application(s)   peptide synthesis: suitable
mp   283 °C (dec.) (lit.)
SMILES string   N[C@@H](CC=C)C(O)=O
InChI   1S/C5H9NO2/c1-2-3-4(6)5(7)8/h2,4H,1,3,6H2,(H,7,8)/t4-/m0/s1
InChI key   WNNNWFKQCKFSDK-BYPYZUCNSA-N

Description

Packaging

1 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?