EMAIL THIS PAGE TO A FRIEND
286036 Aldrich

Harmaline

97%

Synonym: 1-Methyl-7-methoxy-3,4-dihydro-β-carboline, 3,4-Dihydroharmine, 4,9-Dihydro-7-methoxy-1-methyl-3H-pyrido[3,4-b]indole, Dihydroharmine, Harmalol methyl ether, Harmidine, NSC 407285

  • CAS Number 304-21-2

  • Empirical Formula (Hill Notation) C13H14N2O

  • Molecular Weight 214.26

  •  Beilstein Registry Number 207310

  •  EC Number 206-152-6

  •  MDL number MFCD00004955

Purchase

Properties

InChI Key   RERZNCLIYCABFS-UHFFFAOYSA-N
assay   97%
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
  stupéfiantregulated under CDSA - not available from Sigma-Aldrich Canada
mp   232-234 °C (lit.)

Description

Biochem/physiol Actions

CNS stimulant; may act through NMDA receptors

Application

Reactant for preparation of:
• Acylated harmalines via Friedel-Crafts reaction
• Phenacyl derivatives of 1-methyl-7-methoxy-β-carbolines as central nervous system affectors
• Palladium harmaline DMSO chloro complex as anti-tumor agent

Natural product scaffold in preparation of:
• Trypanothione reductase inhibitors†

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UU9800000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?