• USA Home
  • 286079 - Methyl heptadecanoate

EMAIL THIS PAGE TO A FRIEND
286079 Sigma-Aldrich

Methyl heptadecanoate

99%

Synonym: Heptadecanoic acid methyl ester, Methyl margarate

Purchase

Properties

assay   99%
bp   152-153 °C/0.05 mmHg (lit.)
mp   29.8-30.3 °C (lit.)
SMILES string   CCCCCCCCCCCCCCCCC(=O)OC
InChI   1S/C18H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-2/h3-17H2,1-2H3
InChI key   HUEBIMLTDXKIPR-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

H4515 Methyl heptadecanoate, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?