• USA Home
  • 286885 - Normetanephrine hydrochloride

EMAIL THIS PAGE TO A FRIEND
286885 Aldrich

Normetanephrine hydrochloride

98%

Synonym: α-(Aminomethyl)-4-hydroxy-3-methoxybenzyl alcohol hydrochloride

Purchase

Properties

assay   98%
mp   204 °C (dec.)(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?