• USA Home
  • 290289 - 2,5-Difluorobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
290289 Sigma-Aldrich

2,5-Difluorobenzyl alcohol

99%

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Chemical Synthesis, Organic Building Blocks,
assay   99%
refractive index   n20/D 1.49 (lit.)
density   1.29 g/mL at 25 °C (lit.)
SMILES string   OCc1cc(F)ccc1F
InChI   1S/C7H6F2O/c8-6-1-2-7(9)5(3-6)4-10/h1-3,10H,4H2
InChI key   KIKCAPPVFQLOIU-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
201.2 °F
Flash Point(C) 
94 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?