EMAIL THIS PAGE TO A FRIEND
298670 Sigma-Aldrich

DL-Cystine

99%

Synonym: (±)-3,3′-Dithiobis(2-aminopropionicacid)

  • CAS Number 923-32-0

  • Linear Formula HOOCCH(NH2)CH2SSCH2CH(NH2)COOH

  • Molecular Weight 240.30

  •  Beilstein/REAXYS Number 1728095

  •  EC Number 213-094-5

  •  MDL number MFCD00084652

  •  PubChem Substance ID 329753394

Purchase

Properties

InChI Key   LEVWYRKDKASIDU-UHFFFAOYSA-N
assay   99%
mp   227 °C (dec.) (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C8630 DL-Cystine

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?