EMAIL THIS PAGE TO A FRIEND
302856 Aldrich

Hexafluorobenzene

99.9%

  • CAS Number 392-56-3

  • Empirical Formula (Hill Notation) C6F6

  • Molecular Weight 186.05

  •  Beilstein Registry Number 1683438

  •  EC Number 206-876-2

  •  MDL number MFCD00000288

Purchase

Properties

InChI Key   ZQBFAOFFOQMSGJ-UHFFFAOYSA-N
assay   99.9%
refractive index   n20/D 1.377(lit.)
bp   80-82 °C(lit.)
mp   3.7-4.1 °C(lit.)
density   1.612 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
3
RTECS 
DA3050000
Flash Point(F) 
50 °F
Flash Point(C) 
10 °C
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

H8706 Hexafluorobenzene, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?