• USA Home
  • 325910 - Scandium(III) sulfate pentahydrate

EMAIL THIS PAGE TO A FRIEND
325910 Aldrich

Scandium(III) sulfate pentahydrate

99.9%

Synonym: Sulfuric acid scandium(III) salt pentahydrate

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Scandium Catalysts More...
InChI Key   TYCVDJKZMDZUBP-UHFFFAOYSA-H
assay   99.9%

Description

Application

Reactant in thermal decomposition

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?