• USA Home
  • 333808 - Erbium tris[3-(heptafluoropropylhydroxymethylene)-(+)-camphorate]

EMAIL THIS PAGE TO A FRIEND
333808 Sigma-Aldrich

Erbium tris[3-(heptafluoropropylhydroxymethylene)-(+)-camphorate]

98%

Synonym: Er(hfc)3

  • Empirical Formula (Hill Notation) C42H42ErF21O6

  • Molecular Weight 1209.00

  •  MDL number MFCD00198075

  •  PubChem Substance ID 24860184

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Resolution Reagents, Chiral Shift Reagents, NMR - Buffers and Reagents,
assay   98%
optical activity   [α]26/D +118°, c = 1.2 in methylene chloride
mp   142 °C (lit.)
SMILES string   CC1(C)C2CC[C@@]1(C)C(=O)C2=C(O[Er](OC(=C3C4CC[C@@](C)(C3=O)C4(C)C)C(F)(F)C(F)(F)C(F)(F)F)OC(=C5C6CC[C@@](C)(C5=O)C6(C)C)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)F
InChI   1S/3C14H15F7O2.Er/c3*1-10(2)6-4-5-11(10,3)8(22)7(6)9(23)12(15,16)13(17,18)14(19,20)21;/h3*6,23H,4-5H2,1-3H3;/q;;;+3/p-3/b3*9-7+;/t3*6-,11+;/m111./s1
InChI key   NMDRGZSAQXRCOX-PSMAIOHISA-K
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?