• USA Home
  • 369535 - trans-Glutaconic acid

EMAIL THIS PAGE TO A FRIEND
369535 Sigma-Aldrich

trans-Glutaconic acid

technical grade, 90%

  • CAS Number 628-48-8

  • Linear Formula HOOCCH2CH=CHCOOH

  • Molecular Weight 130.10

  •  MDL number MFCD00002784

  •  PubChem Substance ID 24862986

  •  NACRES NA.23

Purchase

Properties

Related Categories Carboxylic Acid Monomers, Materials Science, Monomers, Polymer Science
Quality Level   100
grade   technical grade
assay   90%
mp   137-139 °C (lit.)
SMILES string   [H]\C(CC(O)=O)=C(\[H])C(O)=O
InChI   1S/C5H6O4/c6-4(7)2-1-3-5(8)9/h1-2H,3H2,(H,6,7)(H,8,9)/b2-1+
InChI key   XVOUMQNXTGKGMA-OWOJBTEDSA-N

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?