• USA Home
  • 394777 - Strontium hexaboride

EMAIL THIS PAGE TO A FRIEND
394777 Aldrich

Strontium hexaboride

−325 mesh, 99%

Purchase

Properties

Related Categories Borides, Ceramics, Materials Science, Metal and Ceramic Science
InChI Key   SLJWUVDCYJZRON-UHFFFAOYSA-N
assay   99%
particle size   −325 mesh
density   3.39 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?