• USA Home
  • 397318 - Chlorotriethylgermane

EMAIL THIS PAGE TO A FRIEND
397318 Aldrich

Chlorotriethylgermane

96%

Synonym: Triethylgermanium chloride

Purchase

Properties

Related Categories Chemical Synthesis, Organogermanium, Organometallic Reagents More...
InChI Key   CZKMPDNXOGQMFW-UHFFFAOYSA-N
assay   96%
refractive index   n20/D 1.459(lit.)
bp   173 °C(lit.)
density   1.175 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGIII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?