EMAIL THIS PAGE TO A FRIEND
399574 Aldrich

Hafnium(IV) carbide

−325 mesh

Purchase

Properties

InChI Key   NVDNLVYQHRUYJA-UHFFFAOYSA-N
particle size   −325 mesh
mp   2930 °C(lit.)
density   12.2 g/mL at 25 °C(lit.)

Description

Application

• Used for preparation of ultra-high temperature ceramics
• Reactant in synthesis of hafnium-containing organometallic polymers
• Additive in alloys
• Used in coatings

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

594636 Hafnium(IV) carbide, <1.25 μm particle size

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?