• USA Home
  • 399825 - Thallium(III) fluoride

EMAIL THIS PAGE TO A FRIEND
399825 Aldrich

Thallium(III) fluoride

99%

Purchase

Properties

assay   99%
mp   550 °C (dec.) (lit.)
density   8.36 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1707 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?