• USA Home
  • 401315 - N-(4,5-Dihydro-5-oxo-1-phenyl-1H-pyrazol-3-yl)-9-octadecenamide

EMAIL THIS PAGE TO A FRIEND
401315 Sigma-Aldrich

N-(4,5-Dihydro-5-oxo-1-phenyl-1H-pyrazol-3-yl)-9-octadecenamide

technical grade, 75%

  • CAS Number 74677-80-8

  • Empirical Formula (Hill Notation) C27H41N3O2

  • Molecular Weight 439.63

  •  MDL number MFCD07798661

  •  PubChem Substance ID 24865005

Purchase

Properties

Related Categories Building Blocks, C12 to C37, Chemical Synthesis, Heterocyclic Building Blocks, Pyrazoles More...
grade   technical grade
assay   75%
mp   150-156 °C (lit.)
SMILES string   CCCCCCCC\C=C/CCCCCCCC(=O)NC1=NN(C(=O)C1)c2ccccc2
InChI   1S/C27H41N3O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19-22-26(31)28-25-23-27(32)30(29-25)24-20-17-16-18-21-24/h9-10,16-18,20-21H,2-8,11-15,19,22-23H2,1H3,(H,28,29,31)/b10-9-
InChI key   KIZIRRWZTGNUCT-KTKRTIGZSA-N

Description

General description

N-(4,5-Dihydro-5-oxo-1-phenyl-1H-pyrazol-3-yl)-9-octadecenamide is a heterocyclic building block.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?