• USA Home
  • 404470 - trans-1-Cinnamylpiperazine

EMAIL THIS PAGE TO A FRIEND
404470 Sigma-Aldrich

trans-1-Cinnamylpiperazine

97%

Purchase

Properties

Related Categories Building Blocks, C13, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
Quality Level   100
assay   97%
bp   129 °C/1 mmHg (lit.)
mp   39-44 °C (lit.)
density   0.989 g/mL at 25 °C (lit.)
SMILES string   C1CN(CCN1)C\C=C\c2ccccc2
InChI   1S/C13H18N2/c1-2-5-13(6-3-1)7-4-10-15-11-8-14-9-12-15/h1-7,14H,8-12H2/b7-4+
InChI key   WGEIOMTZIIOUMA-QPJJXVBHSA-N

Description

General description

trans-1-Cinnamylpiperazine is a piperazine derivative.

Application

trans-1-Cinnamylpiperazine was used as a test compound to investigate the in vitro efficacy as an antiprotozoal against Philasterides dicentrarchi. It may be used to synthesize Incentrom A derivatives.

Packaging

10 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?