• USA Home
  • 408042 - Poly[(o-cresyl glycidyl ether)-co-formaldehyde]

EMAIL THIS PAGE TO A FRIEND
408042 Sigma-Aldrich

Poly[(o-cresyl glycidyl ether)-co-formaldehyde]

average Mn ~1,080

Purchase

Properties

Related Categories Engineering Polymers, Formaldehyde Copolymers and Epoxies, Materials Science, Polymer Science, Polymers More...
mol wt   average Mn ~1,080
extent of labeling   4.8 epoxide functionality
  0.445 eq epoxy per 100 g
viscosity   25 cP(lit.)
mp   63-73 °C
  70-75 °C (lit.)
transition temp   Tg 37-39 °C (DSC)
  softening point 68-78 °C (ring and ball)
density   1.12 g/mL at 25 °C (lit.)

Description

Application

For high temperature adhesives, coatings, electrical and laminating products.

Packaging

100 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?