• USA Home
  • 422266 - 4-Fluorobenzamidoxime

EMAIL THIS PAGE TO A FRIEND
422266 Sigma-Aldrich

4-Fluorobenzamidoxime

98%

  • CAS Number 69113-32-2

  • Linear Formula FC6H4C(=NOH)NH2

  • Molecular Weight 154.14

  •  MDL number MFCD00216571

  •  PubChem Substance ID 24866423

  •  NACRES NA.22

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C7-C8, Chemical Synthesis, Fluorinated Building Blocks,
Quality Level   100
assay   98%
mp   92-95 °C (lit.)
SMILES string   N\C(=N/O)c1ccc(F)cc1
InChI   1S/C7H7FN2O/c8-6-3-1-5(2-4-6)7(9)10-11/h1-4,11H,(H2,9,10)
InChI key   OSUPWUQRPLIJKX-UHFFFAOYSA-N

Description

General description

4-Fluorobenzamidoxime is a benzamidoxime derivative containing amidoxime functional group.

Application

4-Fluorobenzamidoxime ((Z)-4-fluoro-N’-hydroxybenzimidamide) may be used in the synthesis of 2-(3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl)benzyl benzoate, by reaction with 2-(chlorocarbonyl)benzyl benzoate.

Packaging

10 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?