• USA Home
  • 422355 - N1-Isopropyldiethylenetriamine

EMAIL THIS PAGE TO A FRIEND
422355 Sigma-Aldrich

N1-Isopropyldiethylenetriamine

technical grade, 75%

  • CAS Number 207399-20-0

  • Linear Formula (CH3)2CHNHCH2CH2NHCH2CH2NH2

  • Molecular Weight 145.25

  •  MDL number MFCD00192242

  •  PubChem Substance ID 24866428

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks, Polyamines More...
grade   technical grade
assay   75%
refractive index   n20/D 1.463 (lit.)
bp   222 °C (lit.)
density   0.88 g/mL at 25 °C (lit.)
SMILES string   CC(C)NCCNCCN
InChI   1S/C7H19N3/c1-7(2)10-6-5-9-4-3-8/h7,9-10H,3-6,8H2,1-2H3
InChI key   BTQYXSSZVKPNLP-UHFFFAOYSA-N

Description

General description

N1-Isopropyldiethylenetriamine is a polyamine. It is an important organic building block used in organic synthesis.

Packaging

5 mL in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2734PSN2 3(8) / PGII
WGK Germany 
WGK 3
Flash Point(F) 
75.2 °F - closed cup
Flash Point(C) 
24 °C - closed cup
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?