EMAIL THIS PAGE TO A FRIEND
449660 Sigma-Aldrich

Barium fluoride

anhydrous, powder, 99.995% trace metals basis

Synonym: Barium difluoride (BaF2 ), Barium difluoride

Purchase

Properties

Related Categories Ba, Barium, Barium Salts, Chemical Synthesis, Crystal Grade Inorganics,
grade   anhydrous
assay   99.995% trace metals basis
form   powder
refractive index   n20/D 1.4744 (lit.)
bp   2260 °C (lit.)
mp   1368 °C (lit.)
density   4.89 g/mL at 25 °C (lit.)
SMILES string   F[Ba]F
InChI   1S/Ba.2FH/h;2*1H/q+2;;/p-2
InChI key   OYLGJCQECKOTOL-UHFFFAOYSA-L

Description

Features and Benefits

Primary glass modifier in fluorozirconate glasses.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1564 6.1 / PGIII
WGK Germany 
1

Single- & multi-channel pipettes from BRAND
Protocols & Articles

Articles

Inorganic Halides for Scintillation

The 1948 discovery by Hofstadter that sodium iodide doped with thallium exhibits extremely high light-yield and conversion efficiency launched the era of modern radiation spectrometry.1 More than hal...
Luke Grocholl
Chemfiles Volume 5 Article 13
Keywords: Absorption, Applications, Materials Science, Methods, Scintillation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?