• USA Home
  • 461776 - 1-Chloro-4-phenylbutane

EMAIL THIS PAGE TO A FRIEND
461776 Sigma-Aldrich

1-Chloro-4-phenylbutane

97%

Purchase

Properties

Related Categories Aryl, Building Blocks, C9 to C12, Chemical Synthesis, Halogenated Hydrocarbons,
Quality Level   100
assay   97%
refractive index   n20/D 1.518 (lit.)
bp   98-102 °C/6 mmHg (lit.)
density   1.026 g/mL at 25 °C (lit.)
SMILES string   ClCCCCc1ccccc1
InChI   1S/C10H13Cl/c11-9-5-4-8-10-6-2-1-3-7-10/h1-3,6-7H,4-5,8-9H2
InChI key   FLLZCZIHURYEQP-UHFFFAOYSA-N

Description

General description

1-Chloro-4-phenylbutane can be synthesized by reacting 4-phenylbutan-1-ol with POCl3 and pyridine.

Application

1-Chloro-4-phenylbutane may be used for the synthesis of 4-(2,4-dinitrophenyl)butan-1-amine. It was used in the synthesis of 1-phenylbutyl-4-phenylpiperidine-4-carbonitrile oxalate.
It may be used in the preparation of tetralin via cyclialkylation with aluminum chloride and ferric chloride.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
219.2 °F - closed cup
Flash Point(C) 
104 °C - closed cup
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?