• USA Home
  • 463000 - Germanium(IV) fluoride

EMAIL THIS PAGE TO A FRIEND
463000 Aldrich

Germanium(IV) fluoride

electronic grade, ≥99.9%

Synonym: Germanium tetrafluoride

Purchase

Properties

Related Categories CVD and ALD Precursors by Metal, Chemical Synthesis, Germanium, Materials Science, Micro/NanoElectronics,
grade   electronic grade
InChI Key   PPMWWXLUCOODDK-UHFFFAOYSA-N
assay   ≥99.9%
mp   −36.5 °C (lit.)
transition temp   sublimation point -36.5 °C
density   2.46 g/mL at -37 °C(lit.)

Description

Application

Employed in studies on room-temperature oxidation of noble metals and the fabrication of SiGe crystallites on glass substrates.

Germanium(IV) fluoride in combination with disilane gas has recently been used to fabricate SiGe crystallites directly on glass substrates. Intense visible light emission from Ge and Si nanocrystals is of particular interest because of potential uses in optoelectronic devices.

General description

Atomic number of base material: 32 Germanium

Packaging

5 g in stainless steel cylinder

Packaged in electropolished stainless steel lecture bottle with a stainless steel valve and CGA 330 outlet.

Recommended products

Monel gas regulators Z562483 or Z405981 (CGA 330) are recommended.

Requires 316 SS gas regulator Z173959 or Z173967.

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3304 - class 2 - PIH-ZONE B - Compressed gas, toxic, corrosive, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

High Purity Metalorganic Precursors for CPV Device Fabrication

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand. Conventional crystalline silicon solar cells have demonstrated remark...
Ravi Kanjolia
Material Matters Volume 5 Article 4
Keywords: Absorption, Chemical vapor deposition, Degradations, Deposition, Mass spectrometry, Nuclear magnetic resonance spectroscopy, Purification, Semiconductor, Solar cells, Spectroscopy, Tissue microarrays, Vaporization

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?