• USA Home
  • 463787 - Nickel(II) oxalate dihydrate

EMAIL THIS PAGE TO A FRIEND
463787 Aldrich

Nickel(II) oxalate dihydrate

99.999% trace metals basis

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Metal and Ceramic Science, Ni, Nickel Salts,
InChI Key   VNYOIRCILMCTHO-UHFFFAOYSA-L
assay   99.999% trace metals basis
form   powder
Featured Industry   Battery Manufacturing
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
Protocols & Articles

Articles

Changing the Landscape of Environmental and Energy Research Through Novel Nanoscale Materials

Energy and environmental catalysts (EECs) form a major group of materials that will continue to drive the growth of industry and research investments. The EEC market is estimated to be between $10−16...
Santanu Chaudhuri
Material Matters, 2012 v7, n4
Keywords: Adsorption, Alternative energy, Applications, Automotive, Catalysis, Diffusion, Environmental, Gene expression, Immobilization, Infrared spectroscopy, Metal organic frameworks, Methods, Molecular probes, Oxidations, Protocols, Reductions, Search, Spectroscopy, Substitutions, Support, Surface Science, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?