• USA Home
  • 465321 - 1-Methyl-3-indoleacetic acid

EMAIL THIS PAGE TO A FRIEND
465321 Sigma-Aldrich

1-Methyl-3-indoleacetic acid

98%

Synonym: 2-(N-Methylindole-3-yl)acetic acid, NSC 78007

  • CAS Number 1912-48-7

  • Empirical Formula (Hill Notation) C11H11NO2

  • Molecular Weight 189.21

  •  MDL number MFCD00130160

  •  PubChem Substance ID 24870268

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C11, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
Quality Level   100
assay   98%
mp   126-128 °C (lit.)
SMILES string   Cn1cc(CC(O)=O)c2ccccc12
InChI   1S/C11H11NO2/c1-12-7-8(6-11(13)14)9-4-2-3-5-10(9)12/h2-5,7H,6H2,1H3,(H,13,14)
InChI key   NAIPEFIYIQFVFC-UHFFFAOYSA-N

Description

General description

1-Methyl-3-indoleacetic acid is an indole derivative. Direct and riboflavin- or Rose-Bengal-sensitized photo-oxidation of 1-methyl-3-indoleacetic acid in aqueous buffer (pH 5 or pH 8) containing 10% methanol has been studied.

Application

• Reactant for enantioselective preparation of α-aryl alkylcarboxylic acids
• Reactant for preparation of PKC inhibitor methylindolylpyridinylmethylpiperidinylindolylpyrroledione for protein kinase C imaging
• Reactant for stereoselective preparation of δ-lactones via Michael addition/lactonization with unsaturated carboxylates
• Reactant for stereoselective preparation of vindorosine, vindoline, and analogs
• Reactant for preparation of (indolyl)pyrrol-2-ones as VEGF-R inhibitors, antiangiogenic agents, and protein kinase inhibitors
• Reactant for synthesis of 3-aryl-5-alkyl-2,5-dihydrofuran-2-ones and their carbanalogues as antifungal agents

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?