EMAIL THIS PAGE TO A FRIEND
491861 Sigma-Aldrich

Benzoic acid-12C7

99.9 atom % 12C

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Purchase

Properties

Related Categories Alphabetical Listings, B, Benzoic Acid and Benzoic Acid Solutions, Essential Chemicals, Research Essentials,
description   13C-depleted
InChI Key   WPYMKLBDIGXBTP-QZNDBYKSSA-N
isotopic purity   99.9 atom % 12C
bp   249 °C (lit.)
mp   121-125 °C (lit.)
mass shift   depleted

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Target organs 
Lungs
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
1
RTECS 
DG0875000
Flash Point(F) 
249.8 °F
Flash Point(C) 
121 °C

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?