EMAIL THIS PAGE TO A FRIEND
510181 Aldrich

Cesium azide

99.99% trace metals basis

Purchase

Properties

Related Categories Azidation/Diazo Transfer, C-X Bond Formation (Non-Halogen), Cesium Salts, Chemical Synthesis, Essential Chemicals,
InChI Key   AYTVLULEEPNWAX-UHFFFAOYSA-N
assay   99.99% trace metals basis
form   powder
mp   310 °C (lit.)
density   3.5 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN 3288 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Organic Azides and Azide Sources

Since the preparation of the first organic azide, phenyl azide, by Peter Griess in 1864 this energy-rich and versatile class of compounds has enjoyed considerable interest. In more recent years, comp...
Matthias Junkers
Aldrich ChemFiles 2008, 8.1, 10.
Keywords: Combinatorial synthesis, Cycloadditions, Hydroazidations, Metathesis, Peptide synthesis

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?